Gene Transcripts

Gene Symbol
Vipr2
Entrez Gene ID
22355
Gene Name
vasoactive intestinal peptide receptor 2
Gene Aliases
VPAC2, VPAC2R, Vip2
Chromosome Location
Chr. 12: 116077726 - 116159604 on Build GRCm38
UniGene ID
Mm.395245
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009511.2 NP_033537.1 7 851
GenBank mRNA AK038743.1 - 7 852
BC138572.1 - 7 749 8861592
D28132.1 - 7 783

View Full Product Details