Gene Transcripts

Gene Symbol
VAT1
Entrez Gene ID
10493
Gene Name
vesicle amine transport 1
Gene Aliases
VATI
Chromosome Location
Chr. 17: 43014605 - 43022442 on Build GRCh38
UniGene ID
Hs.514199
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006373.3 NP_006364.2 5 1035
GenBank mRNA AK097072.1 - 6 862
AK290460.1 - 5 1011
AK298536.1 - 5 745
AK315838.1 - 5 599
BC001913.1 - 5 904
BC008725.2 - 5 959 3162543
BC014279.2 - 5 941 3834206
BC015041.2 - 5 973 3919901
BC090858.1 - 5 973 6528247
BT007369.1 - 5 915
U18009.1 - 5 673
U25779.1 - 5 643

View Full Product Details