Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026113.4 | NP_080389.2 | 6 | 530 | |
GenBank mRNA | AK005261.1 | - | 6 | 529 | |
AK007816.1 | - | 6 | 531 | ||
AK010514.1 | - | 6 | 530 | ||
AK162053.1 | - | 6 | 530 | ||
AK168070.1 | - | 6 | 529 | ||
BC027260.1 | - | 5 | 446 | ||
BC048446.1 | - | 6 | 536 | 6590885 |