Gene Transcripts
- Gene Symbol
- Kirrel3
- Entrez Gene ID
- 67703
- Gene Name
- kin of IRRE like 3 (Drosophila)
- Gene Aliases
- 1500010O20Rik, 2900036G11Rik, NEPH2, SST4, mKIAA1867
- Chromosome Location
- Chr. 9: 34485852 - 35036315 on Build GRCm38
- UniGene ID
- Mm.220710
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001190911.1 |
NP_001177840.1
|
9, 10 |
1502 |
|
NM_001190912.1 |
NP_001177841.1
|
8, 9 |
1424 |
|
NM_001190913.1 |
NP_001177842.1
|
9, 10 |
1502 |
|
NM_001190914.1 |
NP_001177843.1
|
10, 11 |
1603 |
|
NM_026324.3 |
NP_080600.1
|
9, 10 |
1502 |
|
GenBank mRNA |
AK034142.1 |
- |
9, 10 |
1439 |
|
AK038385.1 |
- |
9, 10 |
1253 |
|
AK045373.1 |
- |
9, 10 |
1407 |
|
AK079332.1 |
- |
10, 11 |
1541 |
|
AK171866.1 |
- |
6, 7 |
853 |
|
AY169782.1 |
- |
9, 10 |
1502 |
|
BC063072.1 |
- |
8, 9 |
1219 |
5695003 |
View Full Product Details