Gene Transcripts

Gene Symbol
Tln2
Entrez Gene ID
70549
Gene Name
talin 2
Gene Aliases
5730421P04Rik, AI507121, AI787438, AL118320, mKIAA0320
Chromosome Location
Chr. 9: 67217085 - 67634036 on Build GRCm38
UniGene ID
Mm.33645
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081242.2 NP_001074711.2 31 4107
GenBank mRNA AB093231.1 - 5 649
AF467081.1 - 29 3836
AK029828.1 - 6 800
BC079896.1 - 22 3068 30544295
BC158057.1 - 31 4078 9007357

View Full Product Details