Gene Transcripts

Gene Symbol
Dram1
Entrez Gene ID
71712
Gene Name
DNA-damage regulated autophagy modulator 1
Gene Aliases
1200002N14Rik, Dram
Chromosome Location
Chr. 10: 88322801 - 88364383 on Build GRCm38
UniGene ID
Mm.35450
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027878.2 NP_082154.2 4 577
GenBank mRNA AK004552.1 - 4 577
AK151184.1 - 4 503
BC021433.1 - 4 543 5030012

View Full Product Details