Gene Transcripts

Gene Symbol
Cdc40
Entrez Gene ID
71713
Gene Name
cell division cycle 40
Gene Aliases
1200003H23Rik, EHB3, PRP17
Chromosome Location
Chr. 10: 40831621 - 40883143 on Build GRCm38
UniGene ID
Mm.46063
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027879.2 NP_082155.1 1 177
GenBank mRNA AK004569.1 - 1 144
AK040680.1 - 1 126
AK084140.1 - 1 175
AK135411.1 - 1 144
BC061220.1 - 1 152
BC147503.1 - 1 177 9056386
BC158078.1 - 1 177 9088170

View Full Product Details