Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028102.1 | NP_082378.1 | 2 | 252 | |
GenBank mRNA | AK008529.1 | - | 2 | 238 | |
AK035532.1 | - | 2 | 304 | ||
AK039979.1 | - | 1 | 1269 | ||
AK079311.1 | - | 2 | 300 | ||
AK173010.1 | - | 2 | 204 | ||
AK196094.1 | - | 1 | 168 | ||
BC046229.1 | - | 1 | 172 | ||
BC118962.2 | - | 2 | 487 | 40095026 | |
BY790733.1 | - | 2 | 252 |