Gene Transcripts

Gene Symbol
Isca2
Entrez Gene ID
74316
Gene Name
iron-sulfur cluster assembly 2 homolog (S. cerevisiae)
Gene Aliases
0710001C05Rik, 5730594E03Rik, Hbld1
Chromosome Location
Chr. 12: 84773228 - 84775089 on Build GRCm38
UniGene ID
Mm.25719
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028863.1 NP_083139.1 1 80
GenBank mRNA AK002936.1 - 1 79
AK019988.1 - 1 44
BC022589.1 - 1 85 4037176
BY125913.1 - 1 73

View Full Product Details