Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006407.3 | NP_006398.1 | 3 | 577 | |
GenBank mRNA | AB014765.1 | - | 3 | 483 | |
AB052638.1 | - | 3 | 475 | ||
AB097051.1 | - | 3 | 558 | ||
AF070523.1 | - | 3 | 583 | ||
AF115965.1 | - | 3 | 568 | ||
AF125530.1 | - | 3 | 569 | ||
AF161476.1 | - | 3 | 558 | ||
AK074612.1 | - | 3 | 529 | ||
AK223566.1 | - | 3 | 481 | ||
AK301637.1 | - | 4 | 491 | ||
AK303148.1 | - | 3 | 463 | ||
AK312731.1 | - | 3 | 480 | ||
AY102608.1 | - | 3 | 468 | ||
BC005143.2 | - | 3 | 500 | 3510436 | |
BC020797.1 | - | 3 | 494 | 4134658 |