Gene Transcripts

Gene Symbol
PLK4
Entrez Gene ID
10733
Gene Name
polo like kinase 4
Gene Aliases
MCCRP2, SAK, STK18
Chromosome Location
Chr. 4: 127880861 - 127899224 on Build GRCh38
UniGene ID
Hs.172052
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001190799.1 NP_001177728.1 4 903
NM_001190801.1 NP_001177730.1 5 822
NM_014264.4 NP_055079.3 5 999
XM_005262701.2 XP_005262758.1 5 961
XM_017007662.1 XP_016863151.1 5 1158
XM_017007663.1 XP_016863152.1 5 1159
GenBank mRNA AB006972.1 - 5 913
AK302858.1 - 5 822
AK303399.1 - 4 837
AK314238.1 - 5 967
BC036023.1 - 5 967 5273226
Y13115.1 - 5 865

View Full Product Details