Gene Transcripts

Gene Symbol
RAI2
Entrez Gene ID
10742
Gene Name
retinoic acid induced 2
Gene Aliases
Chromosome Location
Chr. X: 17800049 - 17861346 on Build GRCh38
UniGene ID
Hs.446680
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172732.1 NP_001166203.1 3 1624
NM_001172739.1 NP_001166210.1 3 1968
NM_001172743.1 NP_001166214.1 3 1971
NM_021785.4 NP_068557.3 2 1774
NR_033348.1 3 2047
NR_033349.1 2 1853
XM_006724459.2 XP_006724522.1 4 2123
XM_006724460.1 XP_006724523.1 3 1871
XM_011545439.2 XP_011543741.1 4 2152
XM_011545440.2 XP_011543742.1 4 2155
XM_011545441.2 XP_011543743.1 3 1958
GenBank mRNA AK056214.1 - 3 1867
AK298873.1 - 3 1585
AK314631.1 - 2 1737
BC027937.1 - 3 1927 5201821

View Full Product Details