Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016272.3 | NP_057356.1 | 2 | 1686 | |
XM_005261315.2 | XP_005261372.1 | 2 | 2897 | ||
XM_006724105.3 | XP_006724168.1 | 2 | 2203 | ||
XM_017028539.1 | XP_016884028.1 | 2 | 3979 | ||
GenBank mRNA | AB035207.1 | - | 1 | 1193 | |
AB051450.1 | - | 2 | 1445 | ||
BC038957.1 | - | 2 | 1459 | 5786640 | |
CR456594.1 | - | 2 | 1139 | ||
CR541684.1 | - | 1 | 979 | ||
D64109.1 | - | 1 | 1072 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NR_002936.2 | 1 | 1231 | ||
GenBank mRNA | BC015543.1 | - | 1 | 1251 | |
BC039027.1 | - | 1 | 1269 | ||
BC050562.1 | - | 1 | 1231 |