Gene Transcripts

Gene Symbol
Fam78a
Entrez Gene ID
241303
Gene Name
family with sequence similarity 78, member A
Gene Aliases
A130092J06Rik
Chromosome Location
Chr. 2: 32052166 - 32087436 on Build GRCm38
UniGene ID
Mm.250441
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175511.4 NP_780720.1 1 980
GenBank mRNA AK077626.1 - 1 985
AK079514.1 - 1 981
AK165375.1 - 1348
BC137951.1 - 1 474 8860970
BC137952.1 - 1 474 8860971

View Full Product Details