Gene Transcripts

Gene Symbol
NEK6
Entrez Gene ID
10783
Gene Name
NIMA related kinase 6
Gene Aliases
SID6-1512
Chromosome Location
Chr. 9: 124257606 - 124352442 on Build GRCh38
UniGene ID
Hs.197071
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145001.2 NP_001138473.1 5, 6 666
NM_001166167.1 NP_001159639.1 4, 5 448
NM_001166168.1 NP_001159640.1 4, 5 443
NM_001166169.1 NP_001159641.1 4, 5 452
NM_001166170.1 NP_001159642.1 4, 5 450
NM_001166171.1 NP_001159643.1 5, 6 615
NM_014397.5 NP_055212.2 4, 5 501
XM_005251664.1 XP_005251721.1 4, 5 507
XM_006716936.2 XP_006716999.1 4, 5 512
XM_017014217.1 XP_016869706.1 5, 6 675
GenBank mRNA AB026289.1 - 4, 5 432
AF087909.1 - 4, 5 437
AK294614.1 - 4, 5 452
AK313071.1 - 4, 5 402
BC000101.2 - 4, 5 447 3507262
BC004174.2 - 5, 6 619 2958695
BC004209.2 - 5, 6 619 2958695
BC012761.2 - 4, 5 448 3640103
BM083872.1 - 4, 5 448
BP265230.1 - 4, 5 452
BP279761.1 - 4, 5 443
CF994196.1 - 4, 5 528
CR457091.1 - 3, 4 282
CR542222.1 - 3, 4 282

View Full Product Details