Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001173547.2 | NP_001167018.1 | 7 | 804 | |
NM_001242615.2 | NP_001229544.1 | 7 | 804 | ||
NM_001242616.2 | NP_001229545.1 | 7 | 801 | ||
NM_001290738.1 | NP_001277667.1 | 6 | 723 | ||
NM_181516.6 | NP_852657.1 | 7 | 801 | ||
GenBank mRNA | AK004035.1 | - | 7 | 761 | |
AK075615.1 | - | 7 | 758 | ||
AK076481.1 | - | 7 | 769 | ||
AK160327.1 | - | 7 | 770 | ||
AK170662.1 | - | 7 | 890 | ||
AK178858.1 | - | 4 | 219 | ||
AK183640.1 | - | 2 | 333 | ||
AK188231.1 | - | 5 | 259 | ||
AY231459.1 | - | 7 | 520 | ||
AY231460.1 | - | 7 | 520 | ||
AY258040.1 | - | 7 | 520 | ||
AY258041.1 | - | 7 | 520 | ||
BC015305.1 | - | 7 | 766 | 4221041 | |
CD807936.1 | - | 6 | 669 | ||
EF524069.1 | - | 6 | 400 |