Gene Transcripts

Gene Symbol
Riok3
Entrez Gene ID
66878
Gene Name
RIO kinase 3
Gene Aliases
1200013N13Rik, D18Ertd331e, E130306C24Rik, Sudd
Chromosome Location
Chr. 18: 12128850 - 12157367 on Build GRCm38
UniGene ID
Mm.28551
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024182.4 NP_077144.2 3, 4 378
GenBank mRNA AK004748.1 - 3, 4 470
AK131635.1 - 3, 4 382
AK134327.1 - 3, 4 470
AK147074.1 - 3, 4 379
AK167309.1 - 3, 4 474
AK170882.1 - 3, 4 458
BC033271.1 - 3, 4 416 4016197

View Full Product Details