Gene Transcripts

Gene Symbol
DBF4
Entrez Gene ID
10926
Gene Name
DBF4 zinc finger
Gene Aliases
ASK, CHIF, DBF4A, ZDBF1
Chromosome Location
Chr. 7: 87876229 - 87909541 on Build GRCh38
UniGene ID
Hs.485380
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318060.1 NP_001304989.1 3 798
NM_001318061.1 NP_001304990.1 3 706
NM_001318062.1 NP_001304991.1 3 798
NM_006716.3 NP_006707.1 3 798
GenBank mRNA AB028069.1 - 3 811
AB028070.1 - 3 811
AF160249.1 - 3 510
AF160876.1 - 3 493
AK292569.1 - 3 540
AK302010.1 - 3 427
AK303175.1 - 3 402
BC022811.1 - 3 395
BC036045.1 - 3 604 5273059
BC047693.1 - 3 479 5753567

View Full Product Details