Gene Transcripts

Gene Symbol
MSL3
Entrez Gene ID
10943
Gene Name
male-specific lethal 3 homolog (Drosophila)
Gene Aliases
MSL3L1
Chromosome Location
Chr. X: 11758159 - 11775753 on Build GRCh38
UniGene ID
Hs.655288
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001193270.2 NP_001180199.1 3 326
NM_001282174.1 NP_001269103.1 3 341
NM_006800.3 NP_006791.2 2 311
NM_078628.1 NP_523352.1 3 341
NM_078629.3 NP_523353.2 3 341
XM_005274440.2 XP_005274497.1 3 218
XM_011545442.2 XP_011543744.1 3 295
GenBank mRNA AF117065.1 - 3 341
AK025642.1 - 2 285
AK289780.1 - 3 263
AK294255.1 - 3 211
AK300814.1 - 3 299
AK304419.1 - 2 139
BC031210.1 - 3 274 5313214
BI091178.1 - 3 283
BI560203.1 - 3 329
CT002798.1 - 2 375
DA172776.1 - 3 241
DA788603.1 - 2 313
DA961602.1 - 1 412
DC395802.1 - 3 299

View Full Product Details