Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317182.1 | NP_001304111.1 | 6 | 677 | |
NM_001317183.1 | NP_001304112.1 | 5 | 677 | ||
NM_006838.3 | NP_006829.1 | 6 | 746 | ||
NR_133673.1 | 6 | 746 | |||
XM_005268583.3 | XP_005268640.1 | 6 | 665 | ||
GenBank mRNA | AK055692.1 | - | 468 | ||
AK091730.1 | - | 5 | 553 | ||
AK125296.1 | - | 6 | 1272 | ||
AK300836.1 | - | 6 | 642 | ||
AK315559.1 | - | 6 | 683 | ||
BC013782.1 | - | 6 | 624 | 4340717 | |
U13261.1 | - | 6 | 646 |