Gene Transcripts

Gene Symbol
LILRA1
Entrez Gene ID
11024
Gene Name
leukocyte immunoglobulin like receptor A1
Gene Aliases
CD85I, LIR-6, LIR6
Chromosome Location
Chr. 19: 54593576 - 54602381 on Build GRCh38
UniGene ID
Hs.710507
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278318.1 NP_001265247.1 4 547
NM_001278319.1 NP_001265248.1 3 341
NM_006863.3 NP_006854.1 4 547
NR_103501.1 3 465
NR_103502.1 3 709
NR_103503.1 3 465
GenBank mRNA AF025529.1 - 4 463
AF025530.1 - 4 475
AK290919.1 - 4 491
AK310122.1 - 2 374
AK311226.1 - 3 407
BC093708.1 - 3 321 7939553
BC101514.1 - 3 321 8069020
DA999265.1 - 3 465
EU915602.1 - 3 310

View Full Product Details