Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001100397.1 | NP_001093867.1 | 25 | 2683 | |
NM_001282899.1 | NP_001269828.1 | 25 | 2554 | ||
NM_001282900.1 | NP_001269829.1 | 24 | 2500 | ||
NM_001282901.1 | NP_001269830.1 | 22 | 2288 | ||
NM_007023.3 | NP_008954.2 | 28 | 2924 | ||
XM_005246246.2 | XP_005246303.1 | 27 | 2737 | ||
XM_006712205.3 | XP_006712268.1 | 27 | 2742 | ||
XM_011510505.2 | XP_011508807.1 | 27 | 2771 | ||
XM_017003196.1 | XP_016858685.1 | 28 | 2791 | ||
XM_017003197.1 | XP_016858686.1 | 24 | 2440 | ||
GenBank mRNA | AB027471.1 | - | 28 | 2794 | |
AB075513.1 | - | 12 | 1139 | ||
AB209681.1 | - | 28 | 4978 | ||
AK294433.1 | - | 22 | 2212 | ||
AK294445.1 | - | 25 | 2403 | ||
AK294914.1 | - | 22 | 2241 | ||
AK296340.1 | - | 24 | 2495 | ||
AK304278.1 | - | 25 | 2443 | ||
AK313084.1 | - | 28 | 2793 | ||
AK316014.1 | - | 25 | 2518 | ||
AK316072.1 | - | 25 | 2450 | ||
BC024004.2 | - | 28 | 2763 | 4823935 | |
BC040183.2 | - | 25 | 2684 | 4801092 | |
KJ534924.1 | - | 28 | 2701 | ||
KJ534925.1 | - | 27 | 2683 | ||
KJ534978.1 | - | 28 | 2737 | ||
U78516.1 | - | 28 | 2845 |