Gene Transcripts

Gene Symbol
KERA
Entrez Gene ID
11081
Gene Name
keratocan
Gene Aliases
CNA2, KTN, SLRR2B
Chromosome Location
Chr. 12: 91050491 - 91058354 on Build GRCh38
UniGene ID
Hs.125750
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007035.3 NP_008966.1 2 1353
GenBank mRNA AF063301.1 - 1 742
AF205403.1 - 2 982
AK314990.1 - 2 915
BC032667.2 - 1 743 5541591

View Full Product Details