Gene Transcripts
- Gene Symbol
- Cluap1
- Entrez Gene ID
- 363544
- Gene Name
- clusterin associated protein 1
- Gene Aliases
- Chromosome Location
- Chr. 10: 11847058 - 11884183 on Build Rnor_6.0
- UniGene ID
- Rn.19522
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001014225.1 |
NP_001014247.1
|
6, 7 |
930 |
|
XM_006245832.2 |
XP_006245894.1
|
6, 7 |
693 |
|
XM_008767512.1 |
XP_008765734.1
|
6, 7 |
626 |
|
XM_008767513.1 |
XP_008765735.1
|
6, 7 |
988 |
|
XM_008767514.1 |
XP_008765736.1
|
5, 6 |
530 |
|
XM_008767515.1 |
XP_008765737.1
|
5, 6 |
534 |
|
XM_008767516.1 |
XP_008765738.1
|
5, 6 |
532 |
|
XM_008767517.1 |
XP_008765739.1
|
4, 5 |
395 |
|
XM_008767518.1 |
XP_008765740.1
|
4, 5 |
452 |
|
GenBank mRNA |
BC079022.1 |
- |
6, 7 |
930 |
|
View Full Product Details