Gene Transcripts

Gene Symbol
Elp5
Entrez Gene ID
287446
Gene Name
elongator acetyltransferase complex subunit 5
Gene Aliases
DERP6, Rai12
Chromosome Location
Chr. 10: 56578981 - 56591364 on Build Rnor_6.0
UniGene ID
Rn.9041
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001718.1 NP_001001718.1 4 580
XM_006246639.2 XP_006246701.1 5 774
GenBank mRNA AY623900.1 - 4 580
BC158569.1 - 5 777 9101477

View Full Product Details