Gene Transcripts

Gene Symbol
Cenpm
Entrez Gene ID
315164
Gene Name
centromere protein M
Gene Aliases
RGD1307309
Chromosome Location
Chr. 7: 123464615 - 123476333 on Build Rnor_6.0
UniGene ID
Rn.11654
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130504.1 NP_001123976.1 5, 6 477
XM_008765720.1 XP_008763942.1 4, 5 639

View Full Product Details