Gene Transcripts

Gene Symbol
Copz2
Entrez Gene ID
360611
Gene Name
coatomer protein complex, subunit zeta 2
Gene Aliases
Chromosome Location
Chr. 10: 84718824 - 84731880 on Build Rnor_6.0
UniGene ID
Rn.4257
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108294.1 NP_001101764.1 3 234
GenBank mRNA FQ225734.1 - 3 238

View Full Product Details