Gene Transcripts

Gene Symbol
Bloc1s4
Entrez Gene ID
364183
Gene Name
biogenesis of lysosomal organelles complex-1, subunit 4, cappuccino
Gene Aliases
Cno, RGD1306037
Chromosome Location
Chr. 14: 79013808 - 79015108 on Build Rnor_6.0
UniGene ID
Rn.8060
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100766.1 NP_001094236.1 1 1239
GenBank mRNA BC091265.1 - 1 1213

View Full Product Details