Gene Transcripts
- Gene Symbol
- RNF24
- Entrez Gene ID
- 11237
- Gene Name
- ring finger protein 24
- Gene Aliases
- G1L
- Chromosome Location
- Chr. 20: 3927309 - 4015591 on Build GRCh38
- UniGene ID
- Hs.547576
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001134337.2 |
NP_001127809.1
|
2, 3 |
307 |
|
NM_001134338.2 |
NP_001127810.1
|
3, 4 |
376 |
|
NM_001321749.1 |
NP_001308678.1
|
2, 3 |
543 |
|
NM_007219.4 |
NP_009150.1
|
2, 3 |
599 |
|
XM_011529145.2 |
XP_011527447.1
|
4, 5 |
622 |
|
XM_011529146.2 |
XP_011527448.1
|
3, 4 |
552 |
|
XM_011529147.2 |
XP_011527449.1
|
4, 5 |
630 |
|
XM_017027620.1 |
XP_016883109.1
|
3, 4 |
644 |
|
XM_017027621.1 |
XP_016883110.1
|
3, 4 |
413 |
|
XM_017027622.1 |
XP_016883111.1
|
3, 4 |
488 |
|
GenBank mRNA |
AL079313.1 |
- |
|
15 |
|
AL096778.1 |
- |
2, 3 |
397 |
|
BC000213.2 |
- |
3, 4 |
330 |
2958640 |
BC039584.1 |
- |
2, 3 |
226 |
5214313 |
BT007406.1 |
- |
1, 2 |
145 |
|
CV811405.1 |
- |
2, 3 |
285 |
|
View Full Product Details