Gene Transcripts

Gene Symbol
H1foo
Entrez Gene ID
502875
Gene Name
H1 histone family, member O, oocyte-specific
Gene Aliases
RGD1566236
Chromosome Location
Chr. 4: 147844658 - 147849779 on Build Rnor_6.0
UniGene ID
Rn.218633
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109351.1 NP_001102821.1 2 189

View Full Product Details