Gene Transcripts

Gene Symbol
Klrg2
Entrez Gene ID
689885
Gene Name
killer cell lectin-like receptor subfamily G, member 2
Gene Aliases
Chromosome Location
Chr. 4: 66370325 - 66381334 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_001072378.4 XP_001072378.2 1 1261
XM_006236344.2 XP_006236406.1 1 1262
XM_006236345.2 XP_006236407.1 1 1262

View Full Product Details