Gene Transcripts

Gene Symbol
UGT2A1
Entrez Gene ID
10941
Gene Name
UDP glucuronosyltransferase family 2 member A1 complex locus
Gene Aliases
UDPGT2A1
Chromosome Location
Chr. 4: 69588417 - 69653249 on Build GRCh38
UniGene ID
Hs.225950
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001252274.2 NP_001239203.2 4, 5 1618
NM_001252275.2 NP_001239204.2 4, 5 1120
NM_001301239.1 NP_001288168.1 2, 3 926
NM_006798.4 NP_006789.3 4, 5 1120
GenBank mRNA AJ006054.1 - 3, 4 1067
AK304249.1 - 4, 5 1618
AK314209.1 - 4, 5 1118
BC142640.1 - 2, 3 988

Gene Symbol
UGT2A2
Entrez Gene ID
574537
Gene Name
UDP glucuronosyltransferase family 2 member A2
Gene Aliases
Chromosome Location
Chr. 4: 69588417 - 69639642 on Build GRCh38
UniGene ID
Hs.225950
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105677.2 NP_001099147.2 3, 4 1033
NM_001301233.1 NP_001288162.1 2, 3 901
GenBank mRNA FJ664272.1 - 3, 4 1033
FJ664273.1 - 2, 3 901

View Full Product Details