Gene Transcripts
- Gene Symbol
- PTTG1IP
- Entrez Gene ID
- 754
- Gene Name
- pituitary tumor-transforming 1 interacting protein
- Gene Aliases
- C21orf1, C21orf3, PBF
- Chromosome Location
- Chr. 21: 44849585 - 44873903 on Build GRCh38
- UniGene ID
- Hs.474010
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004339.3 |
NP_004330.1
|
2, 3 |
464 |
|
GenBank mRNA |
AF149785.1 |
- |
2, 3 |
402 |
|
AK094882.1 |
- |
2, 3 |
197 |
|
AK290139.1 |
- |
2, 3 |
217 |
|
AK315626.1 |
- |
2, 3 |
279 |
|
BC000415.2 |
- |
2, 3 |
221 |
2961420 |
BC012858.1 |
- |
2, 3 |
247 |
3857505 |
BC019295.1 |
- |
2, 3 |
211 |
2823814 |
BC020983.1 |
- |
2, 3 |
228 |
3851447 |
BC031097.1 |
- |
2, 3 |
242 |
5288388 |
BC034250.1 |
- |
2, 3 |
236 |
3862830 |
BG721942.1 |
- |
2, 3 |
403 |
|
DC396020.1 |
- |
2, 3 |
465 |
|
Z50022.1 |
- |
2, 3 |
270 |
|
View Full Product Details