Gene Transcripts

Gene Symbol
SLC46A1
Entrez Gene ID
113235
Gene Name
solute carrier family 46 member 1
Gene Aliases
G21, HCP1, PCFT
Chromosome Location
Chr. 17: 28394642 - 28406630 on Build GRCh38
UniGene ID
Hs.446689
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080669.5 NP_542400.2 3 1233
XM_017024110.1 XP_016879599.1 4 1192
GenBank mRNA AK054669.1 - 3 1231
AL832613.2 - 2 541

View Full Product Details