Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001882.3 | NP_001873.2 | 6 | 1039 | |
GenBank mRNA | AK223457.1 | - | 6 | 916 | |
AK307729.1 | - | 6 | 849 | ||
BC018038.1 | - | 6 | 849 | 4812893 | |
CR541666.1 | - | 6 | 759 | ||
X58022.1 | - | 6 | 805 |