Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001109974.2 | NP_001103444.1 | 2 | 747 | |
NM_001166208.1 | NP_001159680.1 | 3 | 1311 | ||
NM_001166209.1 | NP_001159681.1 | 3 | 1244 | ||
NM_007286.5 | NP_009217.3 | 2 | 747 | ||
XM_005268370.1 | XP_005268427.1 | 2 | 690 | ||
XM_005268371.1 | XP_005268428.1 | 2 | 610 | ||
XM_006714755.3 | XP_006714818.1 | 3 | 1299 | ||
XM_011537552.2 | XP_011535854.1 | 3 | 1296 | ||
XM_017009007.1 | XP_016864496.1 | 3 | 1332 | ||
XM_017009008.1 | XP_016864497.1 | 3 | 1271 | ||
GenBank mRNA | AB028952.1 | - | 2 | 701 | |
AF499136.1 | - | 2 | 661 | ||
AF499137.1 | - | 2 | 661 | ||
AK127049.1 | - | 3 | 1244 | ||
AK290178.1 | - | 2 | 730 | ||
AL831818.1 | - | 3 | 1311 | ||
BC142683.1 | - | 2 | 668 | 40148430 | |
BC146665.1 | - | 2 | 714 | 40148077 | |
BC152538.1 | - | 2 | 701 | ||
Y11072.1 | - | 1 | 205 |