Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000258.2 | NP_000249.1 | 5 | 598 | |
GenBank mRNA | AJ709328.1 | - | 2 | 127 | |
AK312044.1 | - | 5 | 559 | ||
BC009790.1 | - | 5 | 563 | 4292684 | |
M24122.1 | - | 5 | 555 | ||
X07373.1 | - | 5 | 505 |