Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000258.2 | NP_000249.1 | 1, 2 | 224 | |
GenBank mRNA | AF174483.1 | - | 1, 2 | 131 | |
AK312044.1 | - | 1, 2 | 185 | ||
BC009790.1 | - | 1, 2 | 189 | 4292684 | |
BP316922.1 | - | 1, 2 | 224 | ||
M24122.1 | - | 1, 2 | 181 | ||
X07373.1 | - | 1, 2 | 131 |