Gene Transcripts

Gene Symbol
NDUFB4
Entrez Gene ID
4710
Gene Name
NADH:ubiquinone oxidoreductase subunit B4
Gene Aliases
B15, CI-B15
Chromosome Location
Chr. 3: 120596281 - 120602411 on Build GRCh38
UniGene ID
Hs.304613
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001168331.1 NP_001161803.1 1 142
NM_004547.5 NP_004538.2 1 142
GenBank mRNA AF044957.1 - 1 71
BC000855.1 - 1 114 3459215
BC070285.1 - 1 81 6714676
BC146885.1 - 1 113 9056813
BC146887.1 - 1 113 9056815
BC146927.1 - 1 113 9056855
BC146932.1 - 1 113 9056860
BF978138.1 - 142

View Full Product Details