Gene Transcripts

Gene Symbol
PEX1
Entrez Gene ID
5189
Gene Name
peroxisomal biogenesis factor 1
Gene Aliases
HMLR1, PBD1A, PBD1B, ZWS, ZWS1
Chromosome Location
Chr. 7: 92487023 - 92528531 on Build GRCh38
UniGene ID
Hs.164682
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000466.2 NP_000457.1 4 472
NM_001282677.1 NP_001269606.1 4 472
NM_001282678.1 NP_001269607.1 4 507
GenBank mRNA AB008112.1 - 4 443
AB052090.1 - 4 376
AB052091.1 - 4 376
AB052092.1 - 4 376
AB052093.1 - 4 376
AB052094.1 - 4 376
AF026086.1 - 4 436
AF030356.1 - 4 380
AK292955.1 - 4 377
AK293757.1 - 4 415
BC035575.1 - 4 414 5498274
DA313282.1 - 4 473
DC370317.1 - 4 483

View Full Product Details