Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_176894.2 | NP_795713.2 | 2 | 911 | |
GenBank mRNA | AF178982.1 | - | 1 | 382 | |
AF295368.1 | - | 1 | 1000 | ||
AF345565.1 | - | 1 | 860 | ||
AF406692.1 | - | 1 | 827 | ||
AK313208.1 | - | 2 | 910 | ||
BC041116.1 | - | 2 | 884 | 5173463 | |
BC066887.1 | - | 1 | 826 | ||
JF810883.1 | - | 2 | 890 |