Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001243224.1 | NP_001230153.1 | 8 | 1049 | |
NM_004584.2 | NP_004575.1 | 11 | 1185 | ||
XM_006718652.3 | XP_006718715.1 | 9 | 1397 | ||
XM_017018097.1 | XP_016873586.1 | 11 | 1129 | ||
XM_017018098.1 | XP_016873587.1 | 9 | 921 | ||
GenBank mRNA | AK315348.1 | - | 11 | 1124 | |
BC014848.1 | - | 11 | 1185 | 4508954 | |
CR536508.1 | - | 11 | 1092 | ||
U53174.1 | - | 11 | 1168 |