Gene Transcripts

Gene Symbol
DMRTA2
Entrez Gene ID
63950
Gene Name
DMRT like family A2
Gene Aliases
Chromosome Location
Chr. 1: 50417550 - 50423447 on Build GRCh38
UniGene ID
Hs.334481
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032110.2 NP_115486.1 3 2673
XM_011541937.2 XP_011540239.1 2 3462

View Full Product Details