Gene Transcripts

Gene Symbol
NEUROD6
Entrez Gene ID
63974
Gene Name
neuronal differentiation 6
Gene Aliases
Atoh2, MATH2, Math-2, NEX1M, Nex1, bHLHa2
Chromosome Location
Chr. 7: 31337461 - 31340924 on Build GRCh38
UniGene ID
Hs.45152
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022728.3 NP_073565.2 2 1063
GenBank mRNA AF063609.1 - 2 866
AF136169.1 - 2 831
AF248954.1 - 2 1063
AK055224.1 - 1 2160
AK055238.1 - 2 1033
AK289619.1 - 2 864
BC035048.2 - 2 865 5261681

View Full Product Details