Gene Transcripts

Gene Symbol
SRP14
Entrez Gene ID
6727
Gene Name
signal recognition particle 14
Gene Aliases
ALURBP
Chromosome Location
Chr. 15: 40035690 - 40039202 on Build GRCh38
UniGene ID
Hs.533732
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001309434.1 NP_001296363.1 3 273
NM_003134.5 NP_003125.3 3 273
GenBank mRNA AB061546.1 - 3 260
AB451391.1 - 3 187
BC035495.2 - 3 229 4298087
BC071716.1 - 3 209 4476669
BC100031.1 - 3 213 2847600
DA338824.1 - 3 273
U07857.1 - 3 208
X73459.1 - 3 206

View Full Product Details