Gene Transcripts

Gene Symbol
SULT2B1
Entrez Gene ID
6820
Gene Name
sulfotransferase family 2B member 1
Gene Aliases
HSST2
Chromosome Location
Chr. 19: 48552172 - 48599427 on Build GRCh38
UniGene ID
Hs.369331
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004605.2 NP_004596.2 1 227
NM_177973.1 NP_814444.1 2 174
GenBank mRNA BC034694.1 - 2 163
U92314.1 - 1 423
U92315.1 - 2 174

View Full Product Details