Gene Transcripts

Gene Symbol
ZNF3
Entrez Gene ID
7551
Gene Name
zinc finger protein 3
Gene Aliases
A8-51, HF.12, KOX25, PP838, Zfp113
Chromosome Location
Chr. 7: 100063847 - 100082548 on Build GRCh38
UniGene ID
Hs.435302
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278284.1 NP_001265213.1 5 642
NM_001278287.1 NP_001265216.1 5 558
NM_001278290.1 NP_001265219.1 4 403
NM_001278291.1 NP_001265220.1 3 272
NM_001278292.1 NP_001265221.1 4 393
NM_001318135.1 NP_001305064.1 5 520
NM_001318136.1 NP_001305065.1 2 552
NM_001318137.1 NP_001305066.1 3 272
NM_017715.3 NP_060185.2 5 524
NM_032924.4 NP_116313.3 5 524
XM_006716111.1 XP_006716174.1 5 578
XM_017012582.1 XP_016868071.1 5 592
XM_017012583.1 XP_016868072.1 3 741
GenBank mRNA AF193057.1 - 5 487
AF217988.1 - 5 488
AK000223.1 - 3 259
AK091984.1 - 2 524
AK297625.1 - 5 486
AK309719.1 - 5 830
BC011887.2 - 5 448 3831842
BC013603.1 - 5 467 3865426
BC025265.1 - 5 457
BQ071498.1 - 5 354
BT006918.1 - 2 89
BX647182.1 - 4 1165
CB988353.1 - 2 552
DA443888.1 - 3 258
DA623743.1 - 4 367
DC384519.1 - 5 524
HY096067.1 - 4 382
X60153.1 - 3 197

View Full Product Details