Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001136040.2 | NP_001129512.1 | 9 | 1483 | |
NM_001142565.1 | NP_001136037.1 | 9 | 1456 | ||
NM_024811.3 | NP_079087.3 | 9 | 1559 | ||
XM_005274298.4 | XP_005274355.1 | 9 | 1542 | ||
XM_005274299.4 | XP_005274356.1 | 9 | 1486 | ||
XM_005274303.4 | XP_005274360.1 | 10 | 1514 | ||
XM_011545257.2 | XP_011543559.1 | 10 | 1368 | ||
XM_011545258.2 | XP_011543560.1 | 9 | 1440 | ||
XM_011545259.2 | XP_011543561.1 | 9 | 1413 | ||
XM_011545260.2 | XP_011543562.1 | 10 | 1560 | ||
XM_011545261.2 | XP_011543563.1 | 10 | 1561 | ||
XM_011545262.2 | XP_011543564.1 | 10 | 1507 | ||
XM_011545263.2 | XP_011543565.1 | 10 | 1506 | ||
XM_017018345.1 | XP_016873834.1 | 10 | 1341 | ||
GenBank mRNA | AJ275970.1 | - | 8 | 1349 | |
AK022591.1 | - | 4 | 751 | ||
AK092037.1 | - | 10 | 1243 | ||
AK096343.1 | - | 9 | 1439 | ||
AK294578.1 | - | 9 | 1412 | ||
AL512759.1 | - | 3 | 338 | ||
AL832546.1 | - | 7 | 1154 | ||
AL832667.1 | - | 9 | 1446 | ||
AL833332.1 | - | 9 | 1412 | ||
BC018135.1 | - | 9 | 1449 | 3913173 | |
BX537888.1 | - | 9 | 1437 |