Gene Transcripts

Gene Symbol
ANKRD26
Entrez Gene ID
22852
Gene Name
ankyrin repeat domain 26
Gene Aliases
THC2, bA145E8.1
Chromosome Location
Chr. 10: 26938195 - 27100498 on Build GRCh38
UniGene ID
Hs.361041
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256053.1 NP_001242982.1 23, 24 2874
NM_014915.2 NP_055730.2 23, 24 2877
XM_006717423.2 XP_006717486.1 24, 25 3963
XM_006717425.3 XP_006717488.1 24, 25 3963
XM_011519416.2 XP_011517718.1 24, 25 3963
XM_017015928.1 XP_016871417.1 24, 25 3963
XM_017015929.1 XP_016871418.1 25, 26 3951
XM_017015930.1 XP_016871419.1 24, 25 3963
XM_017015931.1 XP_016871420.1 24, 25 3963
XM_017015932.1 XP_016871421.1 24, 25 3963
XM_017015933.1 XP_016871422.1 24, 25 3963
GenBank mRNA AB028997.1 - 23, 24 2852
AK128577.1 - 8, 9 1437
BC130453.1 - 23, 24 2818 40146484

View Full Product Details