Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318876.1 | NP_001305805.1 | 4 | 384 | |
NM_203290.3 | NP_976035.1 | 4 | 384 | ||
XM_011515000.2 | XP_011513302.1 | 4 | 325 | ||
GenBank mRNA | AF008442.1 | - | 4 | 310 | |
AF047441.1 | - | 4 | 320 | ||
AK293561.1 | - | 4 | 310 | ||
BC008118.1 | - | 4 | 275 | ||
BC008863.2 | - | 4 | 310 | 4128104 | |
BM828768.1 | - | 4 | 375 |